Learn More
ORS Nasco UVEX STEALTH SAFETY GLS PK/10
Catalog No. NC2676621
Encompass_Preferred
Honeywell part# S3959. Uvex Stealth is the most popular goggle on the market for its all-day comfort and optimum protection. It offers unmatched protection from chemical splash and impact. Uvex Stealth toric lens design is coated with HydroShield AF .and its elastomer body is surrounded by MMT-Multi-Material Technology. Stealth has an easy and economical lens replacement system. Available in OTG and Readers. Rx insert available. Made in the USA.
Catalog No. NC2676621 Supplier ORS Nasco Supplier No. 763S3959
Honeywell part# S3959. Uvex Stealth is the most popular goggle on the market for its all-day comfort and optimum protection. It offers unmatched protection from chemical splash and impact. Uvex Stealth toric lens design is coated with HydroShield AF .and its elastomer body is surrounded by MMT-Multi-Material Technology. Stealth has an easy and economical lens replacement system. Available in OTG and Readers. Rx insert available. Made in the USA.