Learn More
Honeywell Safety Products™ Uvex™ Tectonic Series Safety Eyewear
Catalog No. 19066182
Change view
Click to view available options
Lens Coating:
Hardcoat
Lens Tint:
Amber
Catalog No. Lens Coating Lens Tint
19-066-182 Hardcoat Amber
1 options
This item is not returnable. View return policy
Catalog No. 19-066-182 Supplier Honeywell Safety Products™ Supplier No. XV202
This item is not returnable. View return policy
Weighs less than one ounce (25.2 grams) for lightweight comfort
  • Provides 99.9% UV protection
  • Flexible temples ensures right amount of tension to comfortable place eyewear
  • Comfort-molded nosepiece fits most wearers
  • Wrap-around 9-base curve lens for side protection
  • Available in a wide array of lens tints
  • Anti-scratch hardcoat lens coating for longer wear
  • Uvextra™ anti-fog coating, available in Clear and TSR Gray tints, helps keep lenses clear in humid environments
Compliance
ANSI Z87.1-2010 standard
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Hardcoat
Certifications/Compliance ANSI Z87.1-2010 standard
Lens Tint Amber
Product Type Safety Eyewear
Frame Color Gloss Black
No. per Case 200/Cs.
Temple Style Flexible
Weight (Metric) 25.2 g