Learn More
Honeywell™ Uvex™ Tomcat™ Safety Glasses
Catalog No. 19050915
Change view
Click to view available options
Lens Coating:
Impact-resistant
Lens Tint:
Clear
Gray
SCT-50
Silver Mirror
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint
19-050-915 Impact-resistant SCT-50
19-050-912 Impact-resistant Clear
19-050-914 Impact-resistant Silver Mirror
19-050-913 Impact-resistant Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-050-915 Supplier Honeywell™ Supplier No. S2454
This item is not returnable. View return policy
Sleek, wraparound metal frame design provides a sporty and stylish look
  • Optically corrected 10-base lens design
  • Tight “wrap” style for peripheral protection with maximum visual clarity and quality
  • Flexible rubber temples and adjustable saddle nosepads for custom fit
  • Durable “gunmetal” frame
  • Impact-resistant polycarbonate lenses block 99.9% of harmful UVA and UVB radiation
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meet ANSI Z87.1-2003 standards.
Lens Coating Impact-resistant
Lens Tint SCT-50
Temple Style Rubber
Lens Material Polycarbonate