Learn More
MSC Uvex Turboshield Welding Lens, Green, Shade 3, 9 in High, 0.086 in Thick
Catalog No. 5000508034
Encompass
The Uvex Turboshield Welding Lens is a green face shield window designed for welding applications, offering a Shade 3 protection level. This lens is bright and uncoated, ensuring clear vision for the user. It is specifically for use with Uvex Turboshield systems and is manufactured in the United States.
  • Provides Shade 3 protection
  • Measures 9 in high, 15-7/8 in wide, and 0.086 in (2.20 mm) thick
  • Complies with ANSI Z87.1-2010 and CSA Z94.3 standards
Catalog No. 50-005-08034 Supplier MSC Supplier No. 61455499
The Uvex Turboshield Welding Lens is a green face shield window designed for welding applications, offering a Shade 3 protection level. This lens is bright and uncoated, ensuring clear vision for the user. It is specifically for use with Uvex Turboshield systems and is manufactured in the United States.
  • Provides Shade 3 protection
  • Measures 9 in high, 15-7/8 in wide, and 0.086 in (2.20 mm) thick
  • Complies with ANSI Z87.1-2010 and CSA Z94.3 standards