Learn More
MSC Uvex Welding Window, Green, Shade 3, 0.06 inch Thick, ANSI Z87+
Catalog No. 5000507221
Encompass
This welding window is designed for use with compatible bionic headgear, providing reliable protection. It features a green tint with a shade 3 rating and a thickness of 0.06 inch, meeting ANSI Z87+ standards for safety.
  • Designed for welding applications.
  • Features a bright/uncoated finish.
  • Provides a shade 3 lens.
  • Meets ANSI Z87+ safety standards.
  • Has a thickness of 0.06 "
  • Compatible with bionic headgear.
  • Offers a green tint.
Catalog No. 50-005-07221 Supplier MSC Supplier No. 83715953
This welding window is designed for use with compatible bionic headgear, providing reliable protection. It features a green tint with a shade 3 rating and a thickness of 0.06 inch, meeting ANSI Z87+ standards for safety.
  • Designed for welding applications.
  • Features a bright/uncoated finish.
  • Provides a shade 3 lens.
  • Meets ANSI Z87+ safety standards.
  • Has a thickness of 0.06 "
  • Compatible with bionic headgear.
  • Offers a green tint.