Learn More
Honeywell™ Uvex™ XC™ Safety Glasses Replacement Lenses
Click to view available options
Lens Tint:
Clear
Gray
SCT-Gray
SCT-Low IR
SCT-Reflect 50
Shade 2
Shade 5
Description
Uvex Genesis XC has the same comfort and adjustability features as original Genesis, with 20% more extended wrap-around lens for extra cheek and side coverage. Rx lens insert available. Uvex Genesis XC has an easy and economical lens replacement system.
- PC lens, Hydroshield AF, Dura-streme AF/HC
- Available in a very wide range of lens tints
- 20% more coverage than original Genesis
- Economical lens replacement system
- Meets military Vo ballistic impact requirement (on select tints)
Specifications
Specifications
Lens Type | Uvextreme |
Lens Tint | SCT-Gray |
Product Type | Replacement lens |
For Use With (Equipment) | XC Spectacles |