Learn More
TARGETMOL CHEMICALS INC UVI 3003 10MG
SDP
Catalog No. 502577315
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UVI 3003 is a highly selective antagonist of the retinoid X receptor. UVI 3003 inhibits Xenopus and human RXR(alpha) in Cos7 cells (IC50s 0.22 and 0.24 uM respectively). purity: 99%
Catalog No. 50-257-7315 Supplier TARGETMOL CHEMICALS INC Supplier No. T1720910MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UVI 3003 is a highly selective antagonist of the retinoid X receptor. UVI 3003 inhibits Xenopus and human RXR(alpha) in Cos7 cells (IC50s 0.22 and 0.24 uM respectively). purity: 99%