Learn More
Medchemexpress LLC HY-107500 5mg Medchemexpress, UVI 3003 CAS:847239-17-2 Purity:>98%
SDP
Catalog No. 501968426
Encompass_Preferred
Medchemexpress, HY-107500 5mg UVI 3003 CAS:847239-17-2 UVI 3003 is a highly selective antagonist of retinoid X receptor (RXR), and inhibits xenopus and human RXRα in Cos7 cells, with IC50s of 0.22 and 0.24 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8426 Supplier Medchemexpress LLC Supplier No. HY1075005MG
Medchemexpress, HY-107500 5mg UVI 3003 CAS:847239-17-2 UVI 3003 is a highly selective antagonist of retinoid X receptor (RXR), and inhibits xenopus and human RXRα in Cos7 cells, with IC50s of 0.22 and 0.24 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.