Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVI 3003, Tocris Bioscience™

Catalog No. 330310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-031-0 10 mg
33-035-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-031-0 Supplier R&D Systems Supplier No. 3303/10
This item is not returnable. View return policy

RXR antagonist

RXR antagonist; displays high RXR binding affinity. Does not affect the corepressor interaction capacity of the RARα subunit within the context of the RAR-RXR heterodimer.

Chemical Identifiers

CAS 847239-17-2
Molecular Formula C28H36O4
Molecular Weight (g/mol) 436.592
InChI Key APJSHECCIRQQDV-ZRDIBKRKSA-N
Synonym compound 10e pmid: 19408900, d09ien, 3-4-hydroxy-3-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-pentyloxy-2-naphthalenyl phenyl-2-propenoicacid, e-3-4-hydroxy-3-1,1,4,4-tetramethyl-7-pentoxy-tetralin-6-yl phenyl prop-2-enoic acid, 2e-3-4-hydroxy-3-5,5,8,8-tetramethyl-3-pentyloxy-5,6,7,8-tetrahydronaphthalen-2-yl phenyl prop-2-enoic acid, 3-4-hydroxy-3-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-pentyloxy-2-naphthalenyl phenyl-2-propenoic acid
PubChem CID 44566108
IUPAC Name (E)-3-[4-hydroxy-3-(5,5,8,8-tetramethyl-3-pentoxy-6,7-dihydronaphthalen-2-yl)phenyl]prop-2-enoic acid
SMILES CCCCCOC1=CC2=C(C=C1C3=C(C=CC(=C3)C=CC(=O)O)O)C(CCC2(C)C)(C)C

Specifications

Quantity 10 mg
Formula Weight 436.58
Percent Purity >98%
Chemical Name or Material UVI 3003
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.