Learn More
Glo Germ Company UVL 1003 - 9 LED Focused UV Flashlight, 3 AAA batteries included
SDP
Catalog No. NC0368843
Encompass_Preferred
This powerful new 9 LED focused ultraviolet flash light is light weight and compact featuring an anodized aluminum no-slip grip and a wrist strap. Twist on to inspect hands and surfaces. Expose weaknesses and improve hygiene practices! Requires one AA battery (included).
Catalog No. NC0368843 Supplier Glo Germ Company Supplier No. U1
This powerful new 9 LED focused ultraviolet flash light is light weight and compact featuring an anodized aluminum no-slip grip and a wrist strap. Twist on to inspect hands and surfaces. Expose weaknesses and improve hygiene practices! Requires one AA battery (included).