Learn More
Medipak UVLI Slit-Top Covers 0561, Color- Amber, Standard Size- 8 in x 14 in, 500 Bags
Catalog No. NC3357142
Encompass_Preferred
For 1-liter IV bags (1000 ml). Amber UVLI™ covers provide a convenient, cost-effective, and disposable solution for protecting light-sensitive medications during transportation and administration. The semi-transparency of our Amber UVLI™ makes it easy to read labels and levels underneath without lifting the cover. Our covers and bags exceed the United States Pharmacopoeia Standard blocking over 90% of harmful UV light between 290nm and 450nm, so you can protect with confidence. Slit-top UVLI™ covers are fully sealed on two sides with the top seal having a convenient unsealed portion for even easier hanging.
Catalog No. NC3357142 Supplier Medipak Supplier No. 0561
For 1-liter IV bags (1000 ml). Amber UVLI™ covers provide a convenient, cost-effective, and disposable solution for protecting light-sensitive medications during transportation and administration. The semi-transparency of our Amber UVLI™ makes it easy to read labels and levels underneath without lifting the cover. Our covers and bags exceed the United States Pharmacopoeia Standard blocking over 90% of harmful UV light between 290nm and 450nm, so you can protect with confidence. Slit-top UVLI™ covers are fully sealed on two sides with the top seal having a convenient unsealed portion for even easier hanging.