Learn More
NEOGEN UVM Broth, 10 kg
SDP
Catalog No. 502015188
Encompass_Preferred
UVM Broth is used for the selective enrichment of Listeria spp and is not intended for use in the diagnosis of disease or other conditions in humans.UVM Broth is a modification of the formula described by Donnelly and Baigent. This formula is used for the selective enrichment of Listeria spp. from food samples. Listeria spp. grow over a pH range of 5.0 - 9.6 and survive in food products with pH levels outside these parameters. Listeria spp. are microaerophilic, Gram-positive, asporogenous, non-encapsulated, non-branching, short, motile rods. Motility is pronounced at 20°C. Identification of Listeria is based on successful isolation of the organism, biochemical characterization, and serological confirmation.For more information visit https://www.neogen.com/solutions/Microbiology/uvm-broth
Catalog No. 50-201-5188 Supplier NEOGEN Supplier No. 700002997
UVM Broth is used for the selective enrichment of Listeria spp and is not intended for use in the diagnosis of disease or other conditions in humans.UVM Broth is a modification of the formula described by Donnelly and Baigent. This formula is used for the selective enrichment of Listeria spp. from food samples. Listeria spp. grow over a pH range of 5.0 - 9.6 and survive in food products with pH levels outside these parameters. Listeria spp. are microaerophilic, Gram-positive, asporogenous, non-encapsulated, non-branching, short, motile rods. Motility is pronounced at 20°C. Identification of Listeria is based on successful isolation of the organism, biochemical characterization, and serological confirmation.For more information visit https://www.neogen.com/solutions/Microbiology/uvm-broth