Learn More
NEOGEN UVM Broth, 500 g
SDP
Catalog No. 502015186
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
UVM Broth is used for the selective enrichment of Listeria spp and is not intended for use in the diagnosis of disease or other conditions in humans.UVM Broth is a modification of the formula described by Donnelly and Baigent. This formula is used for the selective enrichment of Listeria spp. from food samples. Listeria spp. grow over a pH range of 5.0 - 9.6 and survive in food products with pH levels outside these parameters. Listeria spp. are microaerophilic, Gram-positive, asporogenous, non-encapsulated, non-branching, short, motile rods. Motility is pronounced at 20°C. Identification of Listeria is based on successful isolation of the organism, biochemical characterization, and serological confirmation.For more information visit https://www.neogen.com/solutions/Microbiology/uvm-broth
Catalog No. 50-201-5186 Supplier NEOGEN Supplier No. 700002995
This item has a minimum qty of 2 per supplier requirements.
UVM Broth is used for the selective enrichment of Listeria spp and is not intended for use in the diagnosis of disease or other conditions in humans.UVM Broth is a modification of the formula described by Donnelly and Baigent. This formula is used for the selective enrichment of Listeria spp. from food samples. Listeria spp. grow over a pH range of 5.0 - 9.6 and survive in food products with pH levels outside these parameters. Listeria spp. are microaerophilic, Gram-positive, asporogenous, non-encapsulated, non-branching, short, motile rods. Motility is pronounced at 20°C. Identification of Listeria is based on successful isolation of the organism, biochemical characterization, and serological confirmation.For more information visit https://www.neogen.com/solutions/Microbiology/uvm-broth