Learn More
Alkali Scientific ASI™ UVMax™ Specialized Plate, PS/Film, Flat Bottom, 384 Well, 20/Cs
SDP
Catalog No. 503027773
Encompass_Preferred
ASI™ UVMax™ is a specialized plate intended for use in lower wavelength. The maximal transmittance is achieved between 260 and 280 nm, ideal for DNA and protein quantification. The plate with low background will enable users to acquire more accurate experimental data. Unlike conventional laboratory cuvettes, ASI™ UVMax™ Plate (96well) allows for high throughput screening process and eliminates the need for expensive and fragile quartz/glass-type plates. ▪ Plate type▪ Developed for determining protein and/or nucleic acid concentrations▪ Certified for low background and consistent performance at 260 and 280 nm▪ Flat type bottom▪ Uniform surface homogeneity▪ Lid not included▪ Alphanumeric labelling
Catalog No. 50-302-7773 Supplier Alkali Scientific Supplier No. UVM384
Add to Cart
Add to Cart
ASI™ UVMax™ is a specialized plate intended for use in lower wavelength. The maximal transmittance is achieved between 260 and 280 nm, ideal for DNA and protein quantification. The plate with low background will enable users to acquire more accurate experimental data. Unlike conventional laboratory cuvettes, ASI™ UVMax™ Plate (96well) allows for high throughput screening process and eliminates the need for expensive and fragile quartz/glass-type plates. ▪ Plate type▪ Developed for determining protein and/or nucleic acid concentrations▪ Certified for low background and consistent performance at 260 and 280 nm▪ Flat type bottom▪ Uniform surface homogeneity▪ Lid not included▪ Alphanumeric labelling