Learn More
UVP 2UV and 3UV Benchtop Transilluminators
Catalog No. UVP95022302 Shop All UVP Inc. Products
Change view
Click to view available options
Model:
LMs-20E
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
UVP95022302 -
UVP95022002 LMs-20E
UVP95022402 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. UVP95022302 Supplier UVP Supplier No. 95022302
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Up to three wavelengths available in one compact, lightweight unit
  • 8-watt models provide economical alternative to High Performance UV Transilluminators
  • All models include hinged UV blocking cover with adjustable height and UVG long-life filter
Two single-intensity options
  • 3UV models offer selection of longwave UV for extended viewing of gels without photonicking; midrange with increased fluorescence photodocumentation and nucleic acid visualization; shortwave for sample irradiation
  • 2UV models provide longwave and midrange UV
Order Info
2UV Dimensions (including cover): 9 1/2L x 13 1/4W x 4 3/4 in.H (24 x 33.7 x 12.1cm); 3UV Dimensions (including cover): 11L x 14W x 6 in.H (28 x 35.6 x 15.2cm)
Certifications
115V models are ETL, ETL/c certified; 230V1 models are CE certified
1International model.