Learn More
UVP 2UV and 3UV Benchtop Transilluminators
Catalog No. UVP95022402 Shop All UVP Inc. Products
Change view
Click to view available options
Model:
LMs-20E
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
UVP95022402 -
UVP95022302 -
UVP95022002 LMs-20E
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. UVP95022402 Supplier UVP Supplier No. 95022402
Up to three wavelengths available in one compact, lightweight unit
  • 8-watt models provide economical alternative to High Performance UV Transilluminators
  • All models include hinged UV blocking cover with adjustable height and UVG long-life filter
Two single-intensity options
  • 3UV models offer selection of longwave UV for extended viewing of gels without photonicking; midrange with increased fluorescence photodocumentation and nucleic acid visualization; shortwave for sample irradiation
  • 2UV models provide longwave and midrange UV
Order Info
2UV Dimensions (including cover): 9 1/2L x 13 1/4W x 4 3/4 in.H (24 x 33.7 x 12.1cm); 3UV Dimensions (including cover): 11L x 14W x 6 in.H (28 x 35.6 x 15.2cm)
Certifications
115V models are ETL, ETL/c certified; 230V1 models are CE certified
1International model.