Learn More
Andwin Scientific UVP BLACKRAY LAMP 115V
SDP
Catalog No. NC9603820
Encompass_Preferred
The 115V UVP lamp features a lightweight and easy-to-handle structure. It is also impact-resistant and can combat fading, peeling, or cracking as a result of its vulnerability to heat or chemicals. This product has a unique sliding wavelength box that permits uses to use either the longwave or shortwave UV ranges either separately or simultaneously. This product is sold individually.
Catalog No. NC9603820 Supplier Andwin Scientific Supplier No. 95002112
The 115V UVP lamp features a lightweight and easy-to-handle structure. It is also impact-resistant and can combat fading, peeling, or cracking as a result of its vulnerability to heat or chemicals. This product has a unique sliding wavelength box that permits uses to use either the longwave or shortwave UV ranges either separately or simultaneously. This product is sold individually.