Learn More
UVP Lamp Funnel for Blak-Ray™ B-100AP High-Intensity UV Inspection Lamps
Catalog No. UVP19012601 Shop All UVP Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Blak-Ray Model B-100AP, B-100AP/R, B-100SP, B-100YP High Intensity UV Lamp
Catalog No. For Use With (Equipment)
UVP19012601 Blak-Ray Model B-100AP, B-100AP/R, B-100SP, B-100YP High Intensity UV Lamp
1 options
Catalog No. UVP19012601 Supplier UVP Supplier No. 19012601
Accessory for B-100AP High-Intensity UV Inspection Lamps
Lamp funnel for use with B-100AP, B-100AP/R, B-100SP high intensity UV inspection lamps.
Specifications
For Use With (Application) Directs UV source to specific area
For Use With (Equipment) Blak-Ray Model B-100AP, B-100AP/R, B-100SP, B-100YP High Intensity UV Lamp
Material Metal
Packaging Quantity 1 Ea.
Product Type Lamp Funnel