Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVP Blak-Ray™ B-100Y and B-100YP UV Inspection Lamps

Catalog No. UVP95012703 Shop All UVP Inc. Products
Change view
Click to view available options
Case Material:
Cool-Touch™ Housing
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Case Material
UVP95012703 Cool-Touch™ Housing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. UVP95012703 Supplier UVP Supplier No. 95012703

missing translation for 'validMainframeMsg'

High intensity lamps to supply brilliant UV irradiance for optimum fluorescence

  • Features yellow filter to block wavelengths shorter than 500nm
  • Produces three strong mercury lines at 543, 574 and 576nm
  • Particles down to only 10 microns are routinely visible
  • Lamp handle fits into transformer base allowing lamp head to rotate 360°

Model B-100Y:

  • Scratch-resistant, powder painted aluminum housing

Model B-100YP:

  • Cool-touch™ plastic housing

Semiconductor wafer inspection, fine surface particle detection

Order Info

Additional filters available (See Cat. No. UVP38-0009-02).

Specifications

Light Source UV
Voltage 115 V
Wattage 100 W
Case Material Cool-Touch™ Housing
For Use With (Application) Fluorescence inspections, non destructive testing, leak detection, readmission
Packaging Quantity 1 Ea.
Product Type UV Wafer inspection Lamp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.