Description
- Lamp features a high intensity 254nm UV grid
- UVP's special UVG shortwave filter reduces solarization, prolonging the filter life
- Lamp mounts securely in its transformer base or is removable for movement around your work area
- Lamp manufactured with rugged, scratch-resistant plastic
- A snap-on filter assembly can be easily attached to convert this lamp for fluorescent analysis
Bacteria destruction, Sterilization, Photochemistry/Phosphorescence, Lapidary, Fluorescent analysis
Specifications
Specifications
| For Use With (Equipment) | Model UVGD-68 Mineralight Display Lamp |
| Packaging Quantity | 1 Ea. |
| Product Type | Shortwave UV Grid |