Learn More
UVP HC-3000 HybriCycler Hybridization Oven
Catalog No. UVP95032901 Shop All UVP Inc. Products
Change view
Click to view available options
Hybricycler Hybridization Oven
Catalog No.
UVP95032901 Hybricycler Hybridization Oven
1 options
Catalog No. UVP95032901 Supplier UVP Supplier No. 95032901
Clear chamber for 360° visibility
Compact size saves valuable benchtop space—measures only 20D × 10.5W × 16.25 in.H.
•    Carousel constructed of clear 0.38 in.-thick beta-blocking acrylic and polycarbonate; rotates at 11rpm for consistent saturation of samples
•    Touch-sensitive keypad and microprocessor ensure accurate temperature output from ambient to 80°C; readout displays on LED
•    Drawer-style tray under rocker wheel contains spill up to 300mL and pulls out for easy cleanup of spilled media
•    Holds up to eight 35 × 150mm, four 35 × 300mm, eight 50mL bottles, or one 70 × 300mm bottom (with carousel removed)