Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVP Mineralight High-Powered UV Grid Lamp

Catalog No. UVP94001205 Shop All UVP Inc. Products
Change view
Click to view available options
Wavelength:
254 nm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength
UVP94001205 254 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. UVP94001205 Supplier UVP Supplier No. 94001205
Only null left
Add to Cart
Add to Cart

Lamps have grid design, which is a highly uniform 254nm UV source of high-intensity

  • Lamp features a high intensity 254nm UV grid
  • UVP's special UVG shortwave filter reduces solarization, prolonging the filter life
  • Lamp mounts securely in its transformer base or is removable for movement around your work area
  • Lamp manufactured with rugged, scratch-resistant plastic
  • A snap-on filter assembly can be easily attached to convert this lamp for fluorescent analysis

Bacteria destruction, Sterilization, Photochemistry/Phosphorescence, Lapidary, Fluorescent analysis

Specifications

Voltage 115 V
Wavelength 254 nm
Case Material Plastic
Dimensions (L x W x H) 16 x 7.8 x 7.8 in. (406 x 198 x 198 mm)
For Use With (Application) Bacteria destruction, Sterilization, Photochemistry/Phosphorescence, Lapidary, Fluorescent analysis
Height (English) 7.8 in.
Height (Metric) 198 mm
Length (English) 16 in.
Length (Metric) 406 mm
Lighting Intensity 1250 mW/cm2 at 6 in. with filter
Model R-52G
Packaging Quantity 1 Ea.
Width (English) 7.8 in.
Width (Metric) 198 mm
Product Type Handheld UV Grid Lamp
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.