Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVP Portable UV Lamp Replacement Tubes

Catalog No. p-168237 Shop All UVP Inc. Products
Change view
Click to view available options
Wavelength:
365 nm
365/284 nm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength
UVP34001501 365/284 nm
UVP34001601 365 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. UVP34001501 Supplier UVP Supplier No. 34001501

Provide a variety of uses including laboratory, research, or criminology applications. UVP™ Portable UV Lamp Replacement Tubes are for use with Model ML-49 Portable UV Lamp with Handle.

Lamps are lightweight for easy handling

  • Durable plastic housing
  • Six watts
  • Models available with shortwave, longwave UV or a combination shortwave/longwave
  • Combination shortwave/longwave version comes with a snap-on wavelength selector for switching between wavelengths

Specifications

Light Source UV-A
Wattage 6 W
Wavelength 365/284 nm
For Use With (Equipment) Tube multiband shortwave / longwave accessory to handheld UV lamps 6W
Packaging Quantity 1 Ea.
Product Type UV Lamp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.