Description
- Easy to install, lightweight
- Sterilized recirculating airflow
- Antimicrobial coated stainless steel reduces bacterial growth
- Access port measures UV intensity with UV sensor
- Easy UV tube replacement
Specifications
Specifications
| Product Type | Air Circulator Module |
| Lighting Type | UV |
| For Use With (Application) | Use with UVP UV Sterilizing PCR Workstation to recirculate UV decontaminated air to reduce surface and airborne contaminants in the PCR workstation chamber |