Learn More
Perkin Elmer US LLC UV/Vis Monofibers
Catalog No. 502104257
Encompass_Preferred
The Transfer-Optic Accessory is a monofibers holder for LAMBDA 10/20/40/800/900 spectrophotometers. It requires a pair of monofibers, either 2- or 10-meters long. Other optrodes are available upon request.
Catalog No. 50-210-4257 Supplier Perkin Elmer US LLC Supplier No. B0196912
Add to Cart
Add to Cart
Request a Quote
The Transfer-Optic Accessory is a monofibers holder for LAMBDA 10/20/40/800/900 spectrophotometers. It requires a pair of monofibers, either 2- or 10-meters long. Other optrodes are available upon request.