Learn More
PATHO3GEN SOLUTIONS UVZone Replacement Lamp Set
Description
- 254 nm UV-C configuration: The station uses four 254 nm UV-C lamps as part of its footwear-treatment process
- 185 nm ozone-generating configuration: The station uses two 185 nm ozone-generating lamps as part of its operating process
- Multiple service options: Select an individual 254 nm lamp, an individual 185 nm lamp, or the complete six-lamp replacement set
- Complete lamp set: Part 05-2552 includes four long 254 nm UV-C lamps and two short 185 nm ozone-generating lamps
- Maintenance interval: Lamps should be replaced after 10,000 hr. of use or annually as directed in the current Instructions for Use
- Qualified installation: Lamp replacement must be performed by trained staff or a PathO3Gen Solutions representative
- Compatibility verification: Confirm the correct part number and station configuration before ordering or installation
Specifications
Specifications
| Technology | 254 nm UV-C, 185 nm Ozone-generating Ultraviolet |
| Product Type | Lamp Set |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.