Learn More
ATCC UWB1.289+BRCA1; Ovarian Carcinoma; Human (Homo sapiens)
Catalog No. 502382222
Encompass
UWB1.289+BRCA1 is a cell line exhibiting epithelial-like morphology that was isolated in 2003 from the ovaries of a 56-year-old female with carcinoma. It can be used for cancer research.
Catalog No. 50-238-2222 Supplier ATCC Supplier No. CRL2946
UWB1.289+BRCA1 is a cell line exhibiting epithelial-like morphology that was isolated in 2003 from the ovaries of a 56-year-old female with carcinoma. It can be used for cancer research.