Learn More
Ushio America, Inc. UXL-75XE XENON 75 WATT BULB
Catalog No. NC9503861
Encompass_Preferred
UXL-75XE Projector Light Bulb. Color/finish: clear . Average lifetime: 400 hours. Lumens (light output): 1000
Catalog No. NC9503861 Supplier Ushio America, Inc. Supplier No. 5000371
UXL-75XE Projector Light Bulb. Color/finish: clear . Average lifetime: 400 hours. Lumens (light output): 1000