Learn More
Aviva Systems Biology Corp UXT Antibody - N-terminal region (ARP51213_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51213_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against UXT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UXT. Alternate names: ART-27, STAP1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53655