Learn More
Medchemexpress LLC Uzansertib (INCB053914) phosphate | 2088852-47-3 | 99.5% | 611.51 g·mol⁻¹ | C26H29F3N5O7P | 10 MG
SDP
Catalog No. 5000248090
Encompass_Preferred
Uzansertib phosphate is the phosphate salt of uzansertib (INCB053914), an orally active, ATP-competitive pan-PIM kinase inhibitor used for preclinical research. It is supplied as a high-purity solid and is intended for in-vitro and in-vivo studies at research institutions.
Catalog No. 50-002-48090 Supplier Medchemexpress LLC Supplier No. HY101870B10MG
Uzansertib phosphate is the phosphate salt of uzansertib (INCB053914), an orally active, ATP-competitive pan-PIM kinase inhibitor used for preclinical research. It is supplied as a high-purity solid and is intended for in-vitro and in-vivo studies at research institutions.