Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uzansertib phosphate | 2088852-47-3 | 98.0% | 611.51 g/mol | C26H29F3N5O7P | 5 MG
SDP

Catalog No. 5000248927
Encompass_Preferred

Uzansertib phosphate is the phosphate salt of uzansertib, an ATP-competitive pan-PIM kinase inhibitor used in preclinical research. It inhibits PIM1, PIM2, and PIM3 and shows antiproliferative activity in hematologic cancer cell lines. Supplied as a solid for research use only.

  • Potent inhibition of PIM1, PIM2, and PIM3 (reported IC50s: PIM1 0.24 nM, PIM2 30 nM, PIM3 0.12 nM).
  • Phosphate salt improves aqueous properties versus the free base.
  • Suitable for in vitro and in vivo studies of PIM kinase signaling.
  • Soluble in DMSO at high concentrations for assay formulation.
  • Typical reported purity ≈98-99.5% depending on supplier.

Catalog No. 50-002-48927 Supplier Medchemexpress LLC Supplier No. HY101870B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uzansertib phosphate is the phosphate salt of uzansertib, an ATP-competitive pan-PIM kinase inhibitor used in preclinical research. It inhibits PIM1, PIM2, and PIM3 and shows antiproliferative activity in hematologic cancer cell lines. Supplied as a solid for research use only.

  • Potent inhibition of PIM1, PIM2, and PIM3 (reported IC50s: PIM1 0.24 nM, PIM2 30 nM, PIM3 0.12 nM).
  • Phosphate salt improves aqueous properties versus the free base.
  • Suitable for in vitro and in vivo studies of PIM kinase signaling.
  • Soluble in DMSO at high concentrations for assay formulation.
  • Typical reported purity ≈98-99.5% depending on supplier.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.