Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uzansertib phosphate | 2088852-47-3 | 99.5% | 611.51 g/mol | C26H29F3N5O7P | 100 MG
SDP

Catalog No. 5000245567
Encompass_Preferred

Uzansertib phosphate is the phosphate salt of uzansertib, an orally active, ATP-competitive pan-PIM kinase inhibitor supplied as a white to off-white solid for research use. The compound is used in biochemical and cellular assays to investigate PIM kinase activity and JAK/STAT signaling.

  • High purity (99.5%) suitable for research applications.
  • Solid powder form, easy to weigh and store.
  • Molecular formula C26H29F3N5O7P and molecular weight 611.51 g/mol.
  • Used in biochemical kinase assays and cellular proliferation studies.
  • Soluble in DMSO for assay preparation.

Catalog No. 50-002-45567 Supplier Medchemexpress LLC Supplier No. HY101870B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uzansertib phosphate is the phosphate salt of uzansertib, an orally active, ATP-competitive pan-PIM kinase inhibitor supplied as a white to off-white solid for research use. The compound is used in biochemical and cellular assays to investigate PIM kinase activity and JAK/STAT signaling.

  • High purity (99.5%) suitable for research applications.
  • Solid powder form, easy to weigh and store.
  • Molecular formula C26H29F3N5O7P and molecular weight 611.51 g/mol.
  • Used in biochemical kinase assays and cellular proliferation studies.
  • Soluble in DMSO for assay preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.