Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uzansertib phosphate | 2088852-47-3 | 99.5% | 611.51 g/mol | C26H29F3N5O7P | 25 MG
SDP

Catalog No. 5000248181
Encompass_Preferred

Uzansertib phosphate is the phosphate salt of Uzansertib (INCB053914), an orally active, ATP-competitive pan-PIM kinase inhibitor used in preclinical research to probe PIM kinase signaling, inhibit proliferation in hematologic cancer cell lines, and reduce tumor growth in animal models.

  • Orally active, ATP-competitive pan-PIM kinase inhibitor.
  • Low-nanomolar activity against PIM1, PIM2, and PIM3 (PIM1 0.24 nM; PIM2 30 nM; PIM3 0.12 nM).
  • High purity solid, about 99.5% by LCMS.
  • Suitable for in vitro and in vivo studies of PIM signaling and downstream phosphorylation (p70S6K/S6, 4E-BP1, BAD).
  • White to off-white solid; molecular weight 611.51 g/mol; formula C26H29F3N5O7P.

Catalog No. 50-002-48181 Supplier Medchemexpress LLC Supplier No. HY101870B25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uzansertib phosphate is the phosphate salt of Uzansertib (INCB053914), an orally active, ATP-competitive pan-PIM kinase inhibitor used in preclinical research to probe PIM kinase signaling, inhibit proliferation in hematologic cancer cell lines, and reduce tumor growth in animal models.

  • Orally active, ATP-competitive pan-PIM kinase inhibitor.
  • Low-nanomolar activity against PIM1, PIM2, and PIM3 (PIM1 0.24 nM; PIM2 30 nM; PIM3 0.12 nM).
  • High purity solid, about 99.5% by LCMS.
  • Suitable for in vitro and in vivo studies of PIM signaling and downstream phosphorylation (p70S6K/S6, 4E-BP1, BAD).
  • White to off-white solid; molecular weight 611.51 g/mol; formula C26H29F3N5O7P.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.