Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uzansertib phosphate | 2088852-47-3 | 99.5% | 611.51 g/mol | C26H29F3N5O7P | 50 MG
SDP

Catalog No. 5000248827
Encompass_Preferred

Uzansertib phosphate is the phosphate salt of uzansertib (INCB053914), an ATP-competitive pan-PIM kinase inhibitor provided for research use. It is supplied as a white to off-white solid with molecular formula C26H29F3N5O7P and molecular weight 611.51 g/mol. Certificate of analysis indicates high chemical purity suitable for in vitro and preclinical experiments.

  • ATP-competitive pan-PIM kinase inhibitor for research and preclinical studies.
  • High chemical purity (99.5%) as reported on the COA.
  • White to off-white solid form, convenient for assay preparation.
  • Molecular formula C26H29F3N5O7P; molecular weight 611.51 g/mol.
  • Supplied in laboratory-scale quantities suitable for experimental workflows.

Catalog No. 50-002-48827 Supplier Medchemexpress LLC Supplier No. HY101870B50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uzansertib phosphate is the phosphate salt of uzansertib (INCB053914), an ATP-competitive pan-PIM kinase inhibitor provided for research use. It is supplied as a white to off-white solid with molecular formula C26H29F3N5O7P and molecular weight 611.51 g/mol. Certificate of analysis indicates high chemical purity suitable for in vitro and preclinical experiments.

  • ATP-competitive pan-PIM kinase inhibitor for research and preclinical studies.
  • High chemical purity (99.5%) as reported on the COA.
  • White to off-white solid form, convenient for assay preparation.
  • Molecular formula C26H29F3N5O7P; molecular weight 611.51 g/mol.
  • Supplied in laboratory-scale quantities suitable for experimental workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.