Learn More
TARGETMOL CHEMICALS INC Uzansertib phosphate 5MG
SDP
Catalog No. 502879514
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Uzansertib phosphate (INCB053914 phosphate) is an orally active, ATP-competitive inhibitor of pan-PIM kinase(PIM1, PIM2, PIM3 with IC50s of 0.24 nM, 30 nM, 0.12 nM , respectively). Purity 99.75%
Catalog No. 50-287-9514 Supplier TARGETMOL CHEMICALS INC Supplier No. T124775MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Uzansertib phosphate (INCB053914 phosphate) is an orally active, ATP-competitive inhibitor of pan-PIM kinase(PIM1, PIM2, PIM3 with IC50s of 0.24 nM, 30 nM, 0.12 nM , respectively). Purity 99.75%