Learn More
Selleck Chemical LLC UZH2 E2911-25MG
SDP
Catalog No. 5000040301
Encompass_Preferred
UZH2 is a potent and selective METTL3 inhibitor with an IC50 value of 5 nM in TR-FRET assay
Catalog No. 50-000-40301 Supplier Selleck Chemical LLC Supplier No. E291125MG
UZH2 is a potent and selective METTL3 inhibitor with an IC50 value of 5 nM in TR-FRET assay