Learn More
Medchemexpress LLC V-0219 hydrochloride | 2922283-73-4 | 99.39% | C20H26ClF3N4O2 | 25 MG
SDP
Catalog No. 5000355909
Encompass_Preferred
V-0219 hydrochloride (Compound 9) is an orally active, positive allosteric modulator (PAM) of the glucagon-like peptide-1 receptor (GLP-1R). It can be used for obesity-associated diabetes research.
  • Orally active, positive allosteric modulator (PAM) of the glucagon-like peptide-1 receptor (GLP-1R)
  • Can be used for obesity-associated diabetes research
  • IC50 and target: GLP-1R
Catalog No. 50-003-55909 Supplier Medchemexpress LLC Supplier No. 25MGHY143312A
V-0219 hydrochloride (Compound 9) is an orally active, positive allosteric modulator (PAM) of the glucagon-like peptide-1 receptor (GLP-1R). It can be used for obesity-associated diabetes research.
  • Orally active, positive allosteric modulator (PAM) of the glucagon-like peptide-1 receptor (GLP-1R)
  • Can be used for obesity-associated diabetes research
  • IC50 and target: GLP-1R