Learn More
Medchemexpress LLC V-0219 hydrochloride | 2922283-73-4 | 99.4% | C20H26ClF3N4O2 | 100 MG
SDP
Catalog No. 5000354082
Encompass_Preferred
V-0219 hydrochloride is an orally active, positive allosteric modulator (PAM) of the glucagon-like peptide-1 receptor (GLP-1R). It can be used for obesity-associated diabetes research.
  • Orally active compound
  • Positive allosteric modulator of the glucagon-like peptide-1 receptor (GLP-1R)
  • Used for obesity-associated diabetes research
Catalog No. 50-003-54082 Supplier Medchemexpress LLC Supplier No. HY143312A100MG
V-0219 hydrochloride is an orally active, positive allosteric modulator (PAM) of the glucagon-like peptide-1 receptor (GLP-1R). It can be used for obesity-associated diabetes research.
  • Orally active compound
  • Positive allosteric modulator of the glucagon-like peptide-1 receptor (GLP-1R)
  • Used for obesity-associated diabetes research