Learn More
Medchemexpress LLC V-0219 hydrochloride | 2922283-73-4 | MFCD35062234 | 99.4% | 446.89 g/mol | C20H26ClF3N4O2 | 5 MG
SDP
Catalog No. 5000222584
Encompass_Preferred
V-0219 hydrochloride is the hydrochloride salt of an orally active positive allosteric modulator (PAM) of the glucagon-like peptide-1 receptor (GLP-1R). It is supplied as a solid research reagent for in vitro and preclinical studies of GLP-1R modulation and metabolic disease models.
  • High purity (99.39%) for reproducible experimental results.
  • Solid form, convenient to weigh and store.
  • Good DMSO solubility (100 mg/mL) for assay preparation.
  • Suitable for in vitro and preclinical screening applications.
  • Storage guidance provided for sealed, moisture-free conditions and solvent storage at -80 °C (long term) or -20 °C (short term).
Catalog No. 50-002-22584 Supplier Medchemexpress LLC Supplier No. HY143312A5MG
V-0219 hydrochloride is the hydrochloride salt of an orally active positive allosteric modulator (PAM) of the glucagon-like peptide-1 receptor (GLP-1R). It is supplied as a solid research reagent for in vitro and preclinical studies of GLP-1R modulation and metabolic disease models.
  • High purity (99.39%) for reproducible experimental results.
  • Solid form, convenient to weigh and store.
  • Good DMSO solubility (100 mg/mL) for assay preparation.
  • Suitable for in vitro and preclinical screening applications.
  • Storage guidance provided for sealed, moisture-free conditions and solvent storage at -80 °C (long term) or -20 °C (short term).