Learn More
Medchemexpress LLC αvβ1 integrin-IN-1 | 1689540-62-2 | MFCD30747846 | 99.5% | 558.65 g·mol⁻1 | C26H34N6O6S | 50 MG
SDP
Catalog No. 5000214767
Encompass_Preferred
αvβ1 integrin-IN-1 is a potent, selective small-molecule inhibitor of the αvβ1 integrin with reported antifibrotic activity. It exhibits sub-nanomolar potency and is provided as a high-purity compound for research applications in vitro and in vivo.
  • Potent inhibition of αvβ1 integrin (IC50 = 0.63 nM).
  • Demonstrated antifibrotic activity in preclinical studies.
  • High purity suitable for research use (reported 99.51%).
  • Molecular weight 558.65 g·mol⁻1 and formula C26H34N6O6S.
  • Supplied in measured quantities for dosing and formulation.
Catalog No. 50-002-14767 Supplier Medchemexpress LLC Supplier No. HY10044550MG
αvβ1 integrin-IN-1 is a potent, selective small-molecule inhibitor of the αvβ1 integrin with reported antifibrotic activity. It exhibits sub-nanomolar potency and is provided as a high-purity compound for research applications in vitro and in vivo.
  • Potent inhibition of αvβ1 integrin (IC50 = 0.63 nM).
  • Demonstrated antifibrotic activity in preclinical studies.
  • High purity suitable for research use (reported 99.51%).
  • Molecular weight 558.65 g·mol⁻1 and formula C26H34N6O6S.
  • Supplied in measured quantities for dosing and formulation.