Learn More
Medchemexpress LLC αvβ1 integrin-IN-1 | 1689540-62-2 | MFCD30747846 | 99.5% | 558.65 g/mol | C26H34N6O6S | 5 MG
SDP
Catalog No. 5000214644
Encompass_Preferred
αvβ1 integrin-IN-1 (Compound C8) is a potent, selective inhibitor of the αvβ1 integrin with an IC50 of 0.63 nM (CAS 1689540-62-2). The compound has reported antifibrotic activity in preclinical studies and is provided for research use.
  • High potency against αvβ1 integrin (IC50 0.63 nM).
  • Reported antifibrotic activity in preclinical models.
  • High purity (99.5%), suitable for biochemical assays.
  • Useful as a chemical probe to study integrin-mediated cell adhesion.
  • Available in research-scale quantities (5 MG).
Catalog No. 50-002-14644 Supplier Medchemexpress LLC Supplier No. HY1004455MG
αvβ1 integrin-IN-1 (Compound C8) is a potent, selective inhibitor of the αvβ1 integrin with an IC50 of 0.63 nM (CAS 1689540-62-2). The compound has reported antifibrotic activity in preclinical studies and is provided for research use.
  • High potency against αvβ1 integrin (IC50 0.63 nM).
  • Reported antifibrotic activity in preclinical models.
  • High purity (99.5%), suitable for biochemical assays.
  • Useful as a chemical probe to study integrin-mediated cell adhesion.
  • Available in research-scale quantities (5 MG).