Learn More
Medchemexpress LLC αvβ1 integrin-IN-1 TFA | 1689540-62-2 | MFCD30747846 | 98.3% | 672.67 g·mol⁻¹ | C28H35F3N6O8S | 10 MG
SDP
Catalog No. 5000215007
Encompass_Preferred
αvβ1 integrin-IN-1 TFA (Compound C8) is a potent, selective small-molecule inhibitor of αvβ1 integrin for research use. It has an IC50 of 0.63 nM and demonstrates antifibrotic effects in mouse models of liver and lung fibrosis. Supplied as a white to off-white solid and as DMSO solutions, it is provided with high purity for biochemical and cellular assays.
  • Potent αvβ1 inhibition with IC50 0.63 nM.
  • High purity (≈98.3%) suitable for research assays.
  • Molecular weight 672.67 g·mol⁻¹.
  • White to off-white solid; available as DMSO solution concentrates.
  • Used to study fibrotic pathways in liver and lung models.
Catalog No. 50-002-15007 Supplier Medchemexpress LLC Supplier No. HY100445A10MG
αvβ1 integrin-IN-1 TFA (Compound C8) is a potent, selective small-molecule inhibitor of αvβ1 integrin for research use. It has an IC50 of 0.63 nM and demonstrates antifibrotic effects in mouse models of liver and lung fibrosis. Supplied as a white to off-white solid and as DMSO solutions, it is provided with high purity for biochemical and cellular assays.
  • Potent αvβ1 inhibition with IC50 0.63 nM.
  • High purity (≈98.3%) suitable for research assays.
  • Molecular weight 672.67 g·mol⁻¹.
  • White to off-white solid; available as DMSO solution concentrates.
  • Used to study fibrotic pathways in liver and lung models.