Learn More
Medchemexpress LLC HY-100445A 5mg Medchemexpress, αvβ1 integrin-IN-1 (TFA) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100445A5MG
Encompass_Preferred
Medchemexpress, HY-100445A 5mg αvβ1 integrin-IN-1 (TFA) CAS:0 αvβ1 integrin-IN-1 TFA (Compound C8) is a potent and selective αvβ1 integrin inhibitor with an IC50 of 0.63 nM. Antifibrotic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9457
 Restricted Item