Learn More
Medchemexpress LLC αvβ1 integrin-IN-1 TFA | 98.3% | 672.67 g·mol⁻1 | C28H35F3N6O8S | 100 MG
SDP
Catalog No. 5000214892
Encompass_Preferred
αvβ1 integrin-IN-1 (Compound C8) is a potent, selective small-molecule inhibitor of the αvβ1 integrin with reported antifibrotic activity. The TFA salt form shows an IC50 of 0.63 nM against αvβ1 and has been used in preclinical studies to reduce fibrotic markers in liver and lung models. Molecular formula C28H35F3N6O8S; molecular weight 672.67 g·mol⁻1.
  • Potent and selective αvβ1 inhibition (IC50 0.63 nM).
  • Demonstrated antifibrotic effects in preclinical models.
  • Suitable for in vitro and in vivo integrin research.
  • Supplied as a TFA salt; analytical data available.
  • Molecular formula C28H35F3N6O8S; molecular weight 672.67 g·mol⁻1.
Catalog No. 50-002-14892 Supplier Medchemexpress LLC Supplier No. HY100445100MG
αvβ1 integrin-IN-1 (Compound C8) is a potent, selective small-molecule inhibitor of the αvβ1 integrin with reported antifibrotic activity. The TFA salt form shows an IC50 of 0.63 nM against αvβ1 and has been used in preclinical studies to reduce fibrotic markers in liver and lung models. Molecular formula C28H35F3N6O8S; molecular weight 672.67 g·mol⁻1.
  • Potent and selective αvβ1 inhibition (IC50 0.63 nM).
  • Demonstrated antifibrotic effects in preclinical models.
  • Suitable for in vitro and in vivo integrin research.
  • Supplied as a TFA salt; analytical data available.
  • Molecular formula C28H35F3N6O8S; molecular weight 672.67 g·mol⁻1.