Learn More
Custom Biogenic Systems V-3000AB Isothermal Freezer, High-Capacity LN2 Storage, -193 °C Average Temperature, 600 lbs Empty
Catalog No. NC3591455
Encompass
The Custom Biogenics Systems V-3000AB Isothermal Freezer offers high-capacity liquid nitrogen storage, ensuring sample integrity and safety with a patented dry, isothermal design. With an average temperature of -193 °C and the ability to hold up to 22,100 vials, it minimizes cross-contamination risk while maximizing accessibility and efficiency thanks to its advanced monitoring system. This freezer is ideal for laboratories needing reliable cryogenic storage solutions.
Catalog No. NC3591455 Supplier Custom Biogenic Systems Supplier No. V3000AB
Add to Cart
Add to Cart
Request a Quote
The Custom Biogenics Systems V-3000AB Isothermal Freezer offers high-capacity liquid nitrogen storage, ensuring sample integrity and safety with a patented dry, isothermal design. With an average temperature of -193 °C and the ability to hold up to 22,100 vials, it minimizes cross-contamination risk while maximizing accessibility and efficiency thanks to its advanced monitoring system. This freezer is ideal for laboratories needing reliable cryogenic storage solutions.