Learn More
Being Scientific V-40 - 40 L/min, 110VAC / 60Hz, Ultimate vacuum level 8 mbar
Catalog No. 5000217564
Encompass_Preferred
THE BEING V Series pumps are characterized in particular by using corrosion-resistant materials such as all-around PTFE for the pump head, FFPM for the valves, and structured diaphragm with a PTFE coating.
Catalog No. 50-002-17564 Supplier Being Scientific Supplier No. BDV15104U
THE BEING V Series pumps are characterized in particular by using corrosion-resistant materials such as all-around PTFE for the pump head, FFPM for the valves, and structured diaphragm with a PTFE coating.