Learn More
Being Scientific V-65 - 65 L/min, 110VAC / 60Hz, Ultimate vacuum level 8 mbar
Catalog No. 5000217565
Encompass_Preferred
THE BEING V Series pumps are characterized in particular by using corrosion-resistant materials such as all-around PTFE for the pump head, FFPM for the valves, and structured diaphragm with a PTFE coating.
Catalog No. 50-002-17565 Supplier Being Scientific Supplier No. BDV15106U
THE BEING V Series pumps are characterized in particular by using corrosion-resistant materials such as all-around PTFE for the pump head, FFPM for the valves, and structured diaphragm with a PTFE coating.