Learn More
MARSHALL SCIENTIFIC Buchi V-800 Vacuum Controller
SDP
Catalog No. NC2591265
Encompass_Preferred
The Buchi V-800 Vacuum Controller is a stand alone valve unit for operation with external vacuum systems, such as the Buchi V-500 vacuum pump which works with the R-200 or R-205 rotovap systems. The V-800 regulates vacuum speed at variable set points and controls constant vacuum. The controller features large LCD display in mbar or torr.
Catalog No. NC2591265 Supplier MARSHALL SCIENTIFIC Supplier No. BUV800
The Buchi V-800 Vacuum Controller is a stand alone valve unit for operation with external vacuum systems, such as the Buchi V-500 vacuum pump which works with the R-200 or R-205 rotovap systems. The V-800 regulates vacuum speed at variable set points and controls constant vacuum. The controller features large LCD display in mbar or torr.