Learn More
MARSHALL SCIENTIFIC Buchi V-850 Vacuum Controller
SDP
Catalog No. NC2591266
Encompass_Preferred
The Buchi V-850 Vacuum Controller is a stand alone valve unit for operation with external vacuum systems, such as the Buchi V-700 vacuum pump which works with the R-210 rotovap system. The easy-to-operate V-850 units are equipped with a single control knob and a large graphic display screen with intuitive menu guidance and six selectable language options. The controller features a solvent database, a timer function for process termination, and a LabVac function for control of laboratory vacuum systems. Units are equipped with a USB port and RS-232/RS-485 interfaces. The integrated aeration valve and the precision pressure sensor are resistant to chemicals, making the controller suitable for numerous applications in the lab.
Catalog No. NC2591266 Supplier MARSHALL SCIENTIFIC Supplier No. BUV850
The Buchi V-850 Vacuum Controller is a stand alone valve unit for operation with external vacuum systems, such as the Buchi V-700 vacuum pump which works with the R-210 rotovap system. The easy-to-operate V-850 units are equipped with a single control knob and a large graphic display screen with intuitive menu guidance and six selectable language options. The controller features a solvent database, a timer function for process termination, and a LabVac function for control of laboratory vacuum systems. Units are equipped with a USB port and RS-232/RS-485 interfaces. The integrated aeration valve and the precision pressure sensor are resistant to chemicals, making the controller suitable for numerous applications in the lab.