Learn More
Medchemexpress LLC HY-112683 10mg Medchemexpress, V-9302 CAS:1855871-76-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11268310MG
Encompass_Preferred
Medchemexpress, HY-112683 10mg V-9302 CAS:1855871-76-9 V-9302 is a competitive antagonist of transmembrane glutamine flux. V-9302 selectively and potently targets the amino acid transporter ASCT2 (SLC1A5). V-9302 inhibits ASCT2-mediated glutamine uptake (IC50=9.6 µM) in HEK-293 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9140